Anti-CAMK2G Rabbit Polyclonal Antibody
Catalog # AVIVARP62969_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:calcium/calmodulin-dependent protein kinase II gamma
- Antigen symbol:CAMK2G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:818
- Antigen synonyms:CAMKG|CAMK|CAMK-II
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CAMK2G (calcium/calmodulin-dependent protein kinase II gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: