Anti-TARS2 Rabbit Polyclonal Antibody
AVIVARP62923_P050
- Antibody type:Primary
- Antigen name:Threonyl-tRNA Synthetase 2
- Antigen symbol:TARS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TARS2 (threonyl-tRNA synthetase 2, mitochondrial (putative))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: