Anti-RRS1 Rabbit Polyclonal Antibody
AVIVARP61752_P050
- Antibody type:Primary
- Antigen name:Ribosome biogenesis regulator homolog
- Antigen symbol:RRS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:23212
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: RRS1 (RRS1 ribosome biogenesis regulator homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat