Anti-ARFGEF1 Rabbit Polyclonal Antibody
Catalog # AVIVARP61685_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ADP-ribosylation factor Guanine Nucleotide-Exchange Factor 1 (Brefeldin A-Inhibited)
- Antigen symbol:ARFGEF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ARFGEF1 (ADP-ribosylation factor guanine nucleotide-exchange factor 1 (brefeldin A-inhibited))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: