Anti-PPP1R2 Rabbit Polyclonal Antibody
AVIVARP61382_P050
- Antibody type:Primary
- Antigen name:protein phosphatase 1, regulatory (inhibitor) subunit 2
- Antigen symbol:PPP1R2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Dog,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5504
- Antigen synonyms:IPP2|IPP-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PPP1R2 (protein phosphatase 1, regulatory (inhibitor) subunit 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig