Anti-CCS Rabbit Polyclonal Antibody
Catalog # AVIVARP61007_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:copper chaperone for superoxide dismutase
- Antigen symbol:CCS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9973
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CCS (copper chaperone for superoxide dismutase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: