Anti-ADORA3 Rabbit Polyclonal Antibody
AVIVARP59956_P050
- Antibody type:Primary
- Antigen name:adenosine A3 receptor
- Antigen symbol:ADORA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:140
- Antigen synonyms:A3AR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ADORA3 (adenosine A3 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: