Anti-CST9 Rabbit Polyclonal Antibody
Catalog # AVIVARP59193_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:cystatin 9 (testatin)
- Antigen symbol:CST9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:128822
- Antigen synonyms:CTES7A|CLM
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CST9 (cystatin 9 (testatin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human