Anti-CDK1 Rabbit Polyclonal Antibody
Catalog # AVIVARP58402_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 1
- Antigen symbol:CDK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:983
- Antigen synonyms:P34CDC2|CDC2|CDC28A
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human