Anti-GPN2 Rabbit Polyclonal Antibody
AVIVARP57104_P050
- Antibody type:Primary
- Antigen name:GPN-loop GTPase 2
- Antigen symbol:GPN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:54707
- Antigen synonyms:ATPBD1B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GPN2 (GPN-loop GTPase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: