Anti-CHTOP Rabbit Polyclonal Antibody
AVIVARP55300_P050
- Antibody type:Primary
- Antigen name:chromatin target of PRMT1
- Antigen symbol:CHTOP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:26097
- Antigen synonyms:C1orf77|pp7704|SRAG-3|SRAG|SRAG-5|FOP|FL-SRAG
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CHTOP (chromatin target of PRMT1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human