Anti-EDDM3B Rabbit Polyclonal Antibody
AVIVARP53756_P050
- Antibody type:Primary
- Antigen name:epididymal protein 3B
- Antigen symbol:EDDM3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:64184
- Antigen synonyms:HE3B|RAM2|HE3-BETA|EP3B|FAM12B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: EDDM3B (epididymal protein 3B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human