Anti-SLC4A5 Rabbit Polyclonal Antibody
Catalog # AVIVARP44021_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 4 (sodium bicarbonate cotransporter), member 5
- Antigen symbol:Solute carrier family 4, member 5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:57835
- Antigen synonyms:NBC4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SLC4A5 (solute carrier family 4 (sodium bicarbonate cotransporter), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: