Anti-CYP1A1 Rabbit Polyclonal Antibody
Catalog # AVIVARP41405_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:cytochrome P450, family 1, subfamily A, polypeptide 1
- Antigen symbol:CYP1A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:1543
- Antigen synonyms:AHRR|CYP1|AHH|P450DX|P450-C|CP11|P1-450
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CYP1A1 (cytochrome P450, family 1, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: