Order Entry
Germany
ContactUsLinkComponent
Anti-RBM12 Rabbit Polyclonal Antibody
Anti-RBM12 Rabbit Polyclonal Antibody
Catalog # AVIVARP40623_P050
Anti-RBM12 Rabbit Polyclonal Antibody
Catalog # AVIVARP40623_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RNA binding motif protein 12
  • Antigen symbol:
    RBM12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    10137
  • Antigen synonyms:
    SWAN|HRIHFB2091
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: RBM12 (RNA binding motif protein 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: