Anti-ZNF529 Rabbit Polyclonal Antibody
AVIVARP39462_P050
- Antibody type:Primary
- Antigen name:zinc finger protein 529
- Antigen symbol:ZNF529
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:57711
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZNF529 (zinc finger protein 529)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Pig