Order Entry
Germany
ContactUsLinkComponent
 
Anti-ZNF12 Rabbit Polyclonal Antibody
  AVIVARP37766_T100
undefined
Anti-ZNF12 Rabbit Polyclonal Antibody
  AVIVARP37766_T100

 

  • Antibody type:
    Primary
  • Antigen name:
    zinc finger protein 12
  • Antigen symbol:
    ZNF12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    7559
  • Antigen synonyms:
    KOX3|ZNF325|GIOT-3|HZF11
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: ZNF12 (zinc finger protein 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: