Anti-ZNF214 Rabbit Polyclonal Antibody
AVIVARP35859_T100
- Antibody type:Primary
- Antigen name:zinc finger protein 214
- Antigen symbol:ZNF214
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:7761
- Antigen synonyms:BAZ1|BAZ-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: ZNF214 (zinc finger protein 214)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: