Anti-GTF3C5 Rabbit Polyclonal Antibody
AVIVARP35843_P050
- Antibody type:Primary
- Antigen name:general transcription factor IIIC, polypeptide 5, 63kDa
- Antigen symbol:GTF3C5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:9328
- Antigen synonyms:TFIIIC63|TFIIICepsilon|TFiiiC2-63
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GTF3C5 (general transcription factor IIIC, polypeptide 5, 63kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: