Anti-TARDBP Rabbit Polyclonal Antibody
AVIVARP35838_P050
- Antibody type:Primary
- Antigen name:TAR DNA binding protein
- Antigen symbol:TARDBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:23435
- Antigen synonyms:ALS10|TDP-43
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TARDBP (TAR DNA binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: