Anti-CHRNA7 Rabbit Polyclonal Antibody
AVIVARP35418_T100
- Antibody type:Primary
- Antigen name:cholinergic receptor, nicotinic, alpha 7 (neuronal)
- Antigen symbol:CHRNA7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:1139
- Antigen synonyms:NACHRA7|CHRNA7-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: CHRNA7 (cholinergic receptor, nicotinic, alpha 7 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: