Anti-GRIK2 Rabbit Polyclonal Antibody
Catalog # AVIVARP35340_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, kainate 2
- Antigen symbol:GRIK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2898
- Antigen synonyms:GLUR6|GLR6|EAA4|GLUK6|MRT6|GluK2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GRIK2 (glutamate receptor, ionotropic, kainate 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: