Order Entry
Germany
ContactUsLinkComponent
 
Anti-NACC1 Rabbit Polyclonal Antibody
  AVIVARP34614_T100
undefined
Anti-NACC1 Rabbit Polyclonal Antibody
  AVIVARP34614_T100

 

  • Antibody type:
    Primary
  • Antigen name:
    nucleus accumbens associated 1, BEN and BTB (POZ) domain containing
  • Antigen symbol:
    NACC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    112939
  • Antigen synonyms:
    BTBD14B|NAC1|NAC-1|BTBD30|BEND8
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: NACC1 (nucleus accumbens associated 1, BEN and BTB (POZ) domain containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat