Anti-CSRP3 Rabbit Polyclonal Antibody
AVIVARP34182_P050
- Antibody type:Primary
- Antigen name:Cysteine and Glycine-rich Protein 3
- Antigen symbol:CSRP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:8048
- Antigen synonyms:LMO4|CLP|MLP|CMH12|CMD1M|CRP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CSRP3 (cysteine and glycine-rich protein 3 (cardiac LIM protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat