Anti-KIF25 Rabbit Polyclonal Antibody
AVIVARP33973_P050
- Antibody type:Primary
- Antigen name:kinesin family member 25
- Antigen symbol:KIF25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3834
- Antigen synonyms:KNSL3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: KIF25 (kinesin family member 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human