Anti-FOXJ2 Rabbit Polyclonal Antibody
AVIVARP32910_T100
- Antibody type:Primary
- Antigen name:Forkhead Box J2
- Antigen symbol:FOXJ2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:55810
- Antigen synonyms:FHX
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: FOXJ2 (forkhead box J2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: