Anti-RAX Rabbit Polyclonal Antibody
Catalog # AVIVARP32164_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Retina and anterior neural fold homeobox
- Antigen symbol:RAX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:30062
- Antigen synonyms:MCOP3|RX
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RAX (retina and anterior neural fold homeobox)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: