Anti-RNF166 Rabbit Polyclonal Antibody
AVIVARP32145_P050
- Antibody type:Primary
- Antigen name:Ring Finger Protein 166
- Antigen symbol:RNF166
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:68718
- Antigen synonyms:1110031E24Rik|Zfp313l|AW555115
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: RNF166 (ring finger protein 166)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: