Anti-HNF4A Rabbit Polyclonal Antibody
AVIVARP31946_P050
- Antibody type:Primary
- Antigen name:hepatocyte nuclear factor 4, alpha
- Antigen symbol:HNF4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3172
- Antigen synonyms:HNF4|HNF4alpha|NR2A1|TCF14|HNF4a8|NR2A21|HNF4a7|TCF|MODY1|MODY|FRTS4|HNF4a9
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: HNF4A (hepatocyte nuclear factor 4, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: