Anti-SEC14L2 Rabbit Polyclonal Antibody
AVIVARP31825_P050
- Antibody type:Primary
- Antigen name:SEC14-like lipid binding 2
- Antigen symbol:SEC14L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:23541
- Antigen synonyms:C22orf6|TAP1|SPF|TAP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SEC14L2 (SEC14-like 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: