Anti-PMAIP1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7074R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Phorbol-12-myristate-13-acetate-induced protein 1
- Antigen symbol:PMAIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:none
- Antigen synonyms:Immediate-early-response protein APR|PMA-induced protein 1|NOXA|Protein Noxa|Phorbol-12-myristate-13-acetate-induced protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PMAIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat