Anti-PPARA Rabbit Polyclonal Antibody
Catalog # BOSSBS-3614R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Peroxisome Proliferator-Activated Receptor Alpha
- Antigen symbol:PPARA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:Nr1c1|PPAR alpha|4933429D07Rik|AW742785|Ppar|PPAR-alpha
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PPARA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat