Anti-CSF3R Rabbit Polyclonal Antibody
Catalog # BOSSBS-2574R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Colony Stimulating Factor 3 receptor (granulocyte)
- Antigen symbol:CSF3R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:GCSFR|CD114
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CSF3R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: