Anti-SLC39A6 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1895R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 39 Member A6
- Antigen symbol:Solute carrier family 39, member 6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:LIV1|ZIP6|Estrogen-regulated protein LIV-1|ZIP-6|LIV-1|Zinc transporter ZIP6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SLC39A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat