Anti-VEGF Rabbit Polyclonal Antibody
BOSSBS-1665R
: Bioss
- Antibody type:Primary
- Antigen name:Vascular endothelial growth factor
- Antigen symbol:VEGF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Sheep,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:VEGFA|VEGF-A|Vascular Permeability Factor|VPF|Vascular Endothelial Growth Factor
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Sheep