Anti-ENPP3 Rabbit polyclonal antibody
Catalog # BOSSBS-1568R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ectonucleotide pyrophosphatase/phosphodiesterase 3
- Antigen symbol:ENPP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ENPP3 (ectonucleotide pyrophosphatase/phosphodiesterase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat