Anti-CK8 Rabbit Polyclonal Antibody
BOSSBS-1106R
: Bioss
- Antibody type:Primary
- Antigen name:Cytokeratin 8
- Antigen symbol:CK8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:cytokeratin 8|Cytokeratin endo A|CYK8|Keratin|KRT8|type II cytoskeletal 8|K8|KRT2-8|CK 8|K2C8|CK8
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: CK8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: