Orthophosphoric acid 10%, EMPROVE® EXPERT Ph. Eur., NF for biopharmaceutical production, SAFC®
Catalog # 4.80951.1000
Supplier: Merck
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:98 g/mol
Density:∼1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
:GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340