Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrochemistry Meters
pH and Multi-Parameter Meters
Accessories for pH Meter, peqMETER 1.14
Accessories for pH Meter, peqMETER 1.14
Accessories for pH Meter, peqMETER 1.14
Supplier:
CONSORT
Certificates
Expand image
Accessories for pH Meter, peqMETER 1.14
Supplier:
CONSORT
Certificates
About this item
Related Information
Documentation
Certificates
Search by Lot Number
CoC_CONSB507_294071406
Documentation