ContactUsLinkComponent
Dishes, roasting, stainless steel
Dishes, roasting, stainless steel
Catalog # CONC5405/300
Supplier:  CONTACTO
undefined
Dishes, roasting, stainless steel
Catalog # CONC5405/300
Supplier:  CONTACTO
Specifications
  • Capacity:
    1500 ml
  • Pk:
    1
  • W×D×H ext.:
    310x200x45 mm
Specifications
About this item
¹⁸/₁₀ stainless steel.
  • Excellent temperature resistance
  • Excellent chemical resistance
  • Easy to clean
Flat edge with extra rounded base and satin polished.