Levelling table, SP Bel-Art
Catalog # BELAH183100000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Levelling table, 203×203×9,5 mm
- Pk:1
Specifications
About this item
This clear acrylic level surface helps when pouring immunological plates, horizontal agarose gels and acrylamide sequencing gels.
- Each leg is adjustable
- Inset levelling bulb below the working surface assists in accurate adjustment