Metal base moulds
Supplier: LEICA HISTOLOGY
Certificates
About this item
Metal base moulds enable tissue to be positioned and embedded in wax when using routine or biopsy cassettes. These re-usable base moulds are made from stainless steel and the internal rounded corners allow for easy removal of the formed wax block.
- Available in packs of 10 or 12
- Can be used on all standard size cassettes