Order Entry
Germany
Orders LinkContactUsLinkComponent
Visor, Visor-Toric

Visor, Visor-Toric

UV-resistant, scratch-resistant, Electrical Arc/short-circuit resistance, PC, 390×200 mm
Catalog # DELPVISORTOIN
Supplier:   DELTA PLUS
Visor, Visor-Toric
Catalog # DELPVISORTOIN
Supplier:  DELTA PLUS
Visor, Visor-Toric
Visor, Visor-Toric

UV-resistant, scratch-resistant, Electrical Arc/short-circuit resistance, PC, 390×200 mm

Price
28,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Clear polycarbonate injected visor with electric arc protection. Suitable for face shield holder VISOR HOLDER and VIROR H (BALBI2 and PICO2 support). Ideal for forest work, building and construction and cutting applications.

  • Universal fixing
  • Protection against large solid particles
  • Easy to use and comfortable

Thickness: 1,9/2,0 mm
Weight: 162 g

Certifications: EN 166, EN 170 (2C-1,2)

Ordering information: The face shield holder is not included and needs to be ordered separately. Please contact your local Avantor office for further details.

Specifications
  • Type:
    UV-resistant, scratch-resistant, Electrical Arc/short-circuit resistance
  • L×W:
    390×200 mm
  • Material:
    PC