Order Entry
Germany
ContactUsLinkComponent
Portable eyewash, Hexafluorine® LPF
Portable eyewash, Hexafluorine® LPF
  PREVLPF
Portable eyewash, Hexafluorine® LPF
  PREVLPF

 

  • Description:
    Portable eyewash pouch, Hexafluorine® LPF, 500 ml
  • Pk:
    500 mL

 

 

This portable eyewash consists of a disposable PVC pouch fitted. Specially adapted for vehicles and mobile teams.

  • Easy to use; without applying any pressure
  • Environmentally friendly

Capacity: 500 ml
Shelf life: 2 years (6 months after opening)