Orders LinkContactUsLinkComponent
Temperature probes
Temperature probes
Supplier:   HUBER
Temperature probes
Supplier:  HUBER
Certificates
Temperature probes
Temperature probes
9 Options Available Below
Product Details & Documents
For external thermo regulation applications.
Product Family Options
Product Information
  • Range
    Length
    Ø×L
    Availability
    Price
  • HUBE10142
    Sensor, G ³/₄, for flow or return
    Sensor, G ³/₄, for flow or return
    −90…+200 °C
    1,5 m
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 565,00 €
    565,00 €
    1 * 1 ST
       565,00 €
  • HUBE6064
    External sensor, PT100
    External sensor, PT100
    −90…+200 °C
    1,5 m
    8×400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 430,00 €
    430,00 €
    1 * 1 ST
       430,00 €
  • HUBE6105
    External sensor, PT100
    External sensor, PT100
    −90…+200 °C
    1,5 m
    6×200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 410,00 €
    410,00 €
    1 * 1 ST
       410,00 €
  • HUBE6138
    External sensor, PT100
    External sensor, PT100
    −90…+200 °C
    1,5 m
    6×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 370,00 €
    370,00 €
    1 * 1 ST
       370,00 €
  • HUBE6205
    External sensor, PT100
    External sensor, PT100
    −90…+200 °C
    1,5 m
    8×170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 530,00 €
    530,00 €
    1 * 1 ST
       530,00 €
  • HUBE6292
    Extension cable, PT100
    Extension cable, PT100
    −90…+200 °C
    3 m
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 265,00 €
    265,00 €
    1 * 1 ST
       265,00 €
  • HUBE6352
    Sensor, M16×1, for flow or return
    Sensor, M16×1, for flow or return
    −90…+200 °C
    1,5 m
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 540,00 €
    540,00 €
    1 * 1 ST
       540,00 €
  • HUBE6353
    Sensor, M16×1, for flow or return double
    Sensor, M16×1, for flow or return double
    −90…+200 °C
    1,5 m
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 600,00 €
    600,00 €
    1 * 1 ST
       600,00 €
  • HUBE6509
    Sensor, M30×1,5, for flow or return
    Sensor, M30×1,5, for flow or return
    −90…+200 °C
    1,5 m
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 630,00 €
    630,00 €
    1 * 1 ST
       630,00 €