VWR® Catalase Reagent
Catalog # 941970ZA
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:30 mL
- Description:Catalase reagent
- Seq:91
Specifications
About this item
The catalase test can differentiate or identify microorganisms by checking their ability to produce catalase, an enzyme that protects microorganisms from toxic oxygen metabolites. Usually only aerobic microorganisms produce catalase.
- A simple test to identify aerotolerant strains Allows identification of Gram-positive cocci Easy to use