Insulin syringes, U100
Supplier: CODAN
Certificates
About this item
These insulin syringes have a Luer slip centric nozzle and an insulin scale.
- Narrow silicone seal
- Dual scale for precise reading
- Plunger stop
- Ergonomic finger grip design
These insulin syringes have a Luer slip centric nozzle and an insulin scale.