Orders LinkContactUsLinkComponent
Capillary columns, HP-1ms
Supplier:   Agilent
Capillary columns, HP-1ms
Supplier:  Agilent
Certificates
Capillary columns, HP-1ms
Capillary columns, HP-1ms
27 Options Available Below
Product Details & Documents
These are excellent general purpose columns.
  • Low bleed characteristics
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Bonded and cross-linked
100% Dimethylpolysiloxane, non-polar, solvent rinsable
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • 558-0246
    on 7" cage
    on 7" cage
    Capillary
    HP-1ms
    0,33 µm
    0,20 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 727,00 €
    727,00 €
    1 * 1 ST
       727,00 €
  • on 7" cage
    Capillary
    HP-1ms
    0,52 µm
    0,32 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 782,00 €
    782,00 €
    1 * 1 ST
       782,00 €
  • on 7" cage
    Capillary
    HP-1ms
    0,50 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 879,00 €
    879,00 €
    1 * 1 ST
       879,00 €
  • on 7" cage
    Capillary
    HP-1ms
    1,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 948,00 €
    948,00 €
    1 * 1 ST
       948,00 €
  • on 7" cage
    Capillary
    HP-1ms
    1,00 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 879,00 €
    879,00 €
    1 * 1 ST
       879,00 €
  • on 7" cage
    Capillary
    HP-1ms
    0,10 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 878,00 €
    878,00 €
    1 * 1 ST
       878,00 €
  • on 7" cage
    Capillary
    HP-1ms
    0,25 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 579,00 €
    579,00 €
    1 * 1 ST
       579,00 €
  • on 7" cage
    Capillary
    HP-1ms
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 948,00 €
    948,00 €
    1 * 1 ST
       948,00 €
  • on 7" cage
    Capillary
    HP-1ms
    0,25 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 630,00 €
    1 630,00 €
    1 * 1 ST
       1 630,00 €
  • on 7" cage
    Capillary
    HP-1ms
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 536,00 €
    536,00 €
    1 * 1 ST
       536,00 €
  • on 7" cage
    Capillary
    HP-1ms
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 878,00 €
    878,00 €
    1 * 1 ST
       878,00 €
  • on 5" cage
    Capillary
    HP-1ms
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 985,00 €
    985,00 €
    1 * 1 ST
       985,00 €
  • on 7" cage
    Capillary
    HP-1ms
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 500,00 €
    1 500,00 €
    1 * 1 ST
       1 500,00 €
  • on 5" cage
    Capillary
    HP-1ms
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 680,00 €
    1 680,00 €
    1 * 1 ST
       1 680,00 €
  • on 5" cage
    Capillary
    HP-1ms
    0,33 µm
    0,20 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 815,00 €
    815,00 €
    1 * 1 ST
       815,00 €
  • on 5" cage
    Capillary
    HP-1ms
    1,00 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 985,00 €
    985,00 €
    1 * 1 ST
       985,00 €
  • on 5" cage
    Capillary
    HP-1ms
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 070,00 €
    1 070,00 €
    1 * 1 ST
       1 070,00 €
  • on 5" cage
    Capillary
    HP-1ms
    0,25 µm
    0,25 mm
    15 m
    Discontinued
  • HEWL19091S-633LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    HP-1ms
    0,50 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • on 7" cage
    Capillary
    HP-1ms
    0,18 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 755,00 €
    755,00 €
    1 * 1 ST
       755,00 €
  • on 5" cage
    Capillary
    HP-1ms
    0,18 µm
    0,18 mm
    20 m
    Discontinued
  • HEWL19091S-677LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    HP-1ms
    0,18 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 010,00 €
    2 010,00 €
    1 * 1 ST
       2 010,00 €
  • HEWL19091S-713LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    HP-1ms
    1,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL19091S-733LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    HP-1ms
    1,00 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 140,00 €
    2 140,00 €
    1 * 1 ST
       2 140,00 €
  • HEWL19091S-833LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    HP-1ms
    0,10 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL19091S-933LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    HP-1ms
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 130,00 €
    2 130,00 €
    1 * 1 ST
       2 130,00 €
  • HEWL29091S-677LTM
    on 5975T LTM Toroid
    on 5975T LTM Toroid
    Capillary
    HP-1ms
    0,18 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL29091S-833LTM
    on 5975T LTM Toroid
    on 5975T LTM Toroid
    Capillary
    HP-1ms
    0,10 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL29091S-931LTM
    on 5975T LTM Toroid
    on 5975T LTM Toroid
    Capillary
    HP-1ms
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST