Orders LinkContactUsLinkComponent
Capillary columns, DB-5
Supplier:   Agilent
Capillary columns, DB-5
Supplier:  Agilent
Certificates
Capillary columns, DB-5
Capillary columns, DB-5
106 Options Available Below
Product Details & Documents
These non-polar columns are used in wide range of applications.
  • Low bleed, bonded and cross-linked
  • Solvent rinsable
  • High temperature limit
(5%-Phenyl)-methylpolysiloxane.
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • HEWL127-5012LTM
    On 7890/6890 LTM II module
    On 7890/6890 LTM II module
    Capillary
    DB-5
    0,10 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 770,00 €
    1 770,00 €
    1 * 1 ST
       1 770,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-5012LTM
  • HEWL127-501ELTM
    On 7890/6890 LTM II module
    On 7890/6890 LTM II module
    Capillary
    DB-5
    0,17 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-501ELTM
  • HEWL127-5013LTM
    On 7890/6890 LTM II module
    On 7890/6890 LTM II module
    Capillary
    DB-5
    0,40 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-5013LTM
  • HEWL12A-5015LTM
    On 7890/6890 LTM II module
    On 7890/6890 LTM II module
    Capillary
    DB-5
    1,20 µm
    0,15 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL12A-5015LTM
  • HEWL121-5022LTM
    On 7890/6890 LTM II module
    On 7890/6890 LTM II module
    Capillary
    DB-5
    0,18 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5022LTM
  • HEWL121-5023LTM
    On 7890/6890 LTM II module
    On 7890/6890 LTM II module
    Capillary
    DB-5
    0,40 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5023LTM
  • HEWL127-5012
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,10 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 518,00 €
    518,00 €
    1 * 1 ST
       518,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-5012
  • HEWL127-5012E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    0,10 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 581,00 €
    581,00 €
    1 * 1 ST
       581,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-5012E
  • HEWL127-501E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    0,17 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 518,00 €
    518,00 €
    1 * 1 ST
       518,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-501E
  • HEWL127-501N
    on 7" cage, high efficiency
    on 7" cage, high efficiency
    Capillary
    DB-5
    0,33 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 518,00 €
    518,00 €
    1 * 1 ST
       518,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-501N
  • HEWL127-5013
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,40 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 518,00 €
    518,00 €
    1 * 1 ST
       518,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-5013
  • HEWL127-5022
    on 7" cage, high efficiency
    on 7" cage, high efficiency
    Capillary
    DB-5
    0,10 µm
    0,10 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 803,00 €
    803,00 €
    1 * 1 ST
       803,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-5022
  • HEWL127-5022E
    on 7" cage, high efficiency
    on 7" cage, high efficiency
    Capillary
    DB-5
    0,10 µm
    0,10 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 901,00 €
    901,00 €
    1 * 1 ST
       901,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-5022E
  • HEWL127-5023
    on 7" cage, high efficiency
    on 7" cage, high efficiency
    Capillary
    DB-5
    0,40 µm
    0,10 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 803,00 €
    803,00 €
    1 * 1 ST
       803,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL127-5023
  • HEWL12A-5015
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,20 µm
    0,15 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 479,00 €
    479,00 €
    1 * 1 ST
       479,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL12A-5015
  • HEWL121-5012
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,18 µm
    0,18 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 415,00 €
    415,00 €
    1 * 1 ST
       415,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5012
  • HEWL121-5012E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    0,18 µm
    0,18 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 466,00 €
    466,00 €
    1 * 1 ST
       466,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5012E
  • HEWL121-5012LTM
    On 7890/6890 LTM II module
    On 7890/6890 LTM II module
    Capillary
    DB-5
    0,18 µm
    0,18 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 620,00 €
    1 620,00 €
    1 * 1 ST
       1 620,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5012LTM
  • HEWL121-5013
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,40 µm
    0,18 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 415,00 €
    415,00 €
    1 * 1 ST
       415,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5013
  • HEWL121-5013LTM
    On 7890/6890 LTM II module
    On 7890/6890 LTM II module
    Capillary
    DB-5
    0,40 µm
    0,18 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 620,00 €
    1 620,00 €
    1 * 1 ST
       1 620,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5013LTM
  • HEWL121-5022
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,18 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 659,00 €
    659,00 €
    1 * 1 ST
       659,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5022
  • HEWL121-5022E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    0,18 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 739,00 €
    739,00 €
    1 * 1 ST
       739,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5022E
  • HEWL121-5023
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,40 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 659,00 €
    659,00 €
    1 * 1 ST
       659,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5023
  • HEWL121-5042
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,18 µm
    0,18 mm
    40 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 220,00 €
    1 220,00 €
    1 * 1 ST
       1 220,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL121-5042
  • HEWL128-5012
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,33 µm
    0,20 mm
    12 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 383,00 €
    383,00 €
    1 * 1 ST
       383,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL128-5012
  • HEWL128-50H7
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,20 µm
    0,20 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 432,00 €
    432,00 €
    1 * 1 ST
       432,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL128-50H7
  • HEWL128-5022
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,33 µm
    0,20 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 650,00 €
    650,00 €
    1 * 1 ST
       650,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL128-5022
  • HEWL128-5052
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,33 µm
    0,20 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 100,00 €
    1 100,00 €
    1 * 1 ST
       1 100,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL128-5052
  • HEWL122-5002LTM
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 660,00 €
    1 660,00 €
    1 * 1 ST
       1 660,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5002LTM
  • HEWL122-5011
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,10 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 459,00 €
    459,00 €
    1 * 1 ST
       459,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5011
  • HEWL122-5012
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 459,00 €
    459,00 €
    1 * 1 ST
       459,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5012
  • HEWL122-5012E
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 515,00 €
    515,00 €
    1 * 1 ST
       515,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5012E
  • HEWL122-5012LTM
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 650,00 €
    1 650,00 €
    1 * 1 ST
       1 650,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5012LTM
  • HEWL122-501E
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,50 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 459,00 €
    459,00 €
    1 * 1 ST
       459,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-501E
  • HEWL122-5013
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 459,00 €
    459,00 €
    1 * 1 ST
       459,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5013
  • HEWL122-5022
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 718,00 €
    718,00 €
    1 * 1 ST
       718,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5022
  • HEWL122-5031
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,10 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 771,00 €
    771,00 €
    1 * 1 ST
       771,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5031
  • HEWL122-5032
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 771,00 €
    771,00 €
    1 * 1 ST
       771,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5032
  • HEWL122-5032E
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 864,00 €
    864,00 €
    1 * 1 ST
       864,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5032E
  • HEWL122-5032LTM
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 000,00 €
    2 000,00 €
    1 * 1 ST
       2 000,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5032LTM
  • HEWL122-503E
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,50 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 771,00 €
    771,00 €
    1 * 1 ST
       771,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-503E
  • HEWL122-503ELTM
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,50 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 000,00 €
    2 000,00 €
    1 * 1 ST
       2 000,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-503ELTM
  • HEWL122-5033
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 771,00 €
    771,00 €
    1 * 1 ST
       771,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5033
  • HEWL122-5033E
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 864,00 €
    864,00 €
    1 * 1 ST
       864,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5033E
  • HEWL122-5033LTM
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 000,00 €
    2 000,00 €
    1 * 1 ST
       2 000,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5033LTM
  • HEWL122-5052
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 300,00 €
    1 300,00 €
    1 * 1 ST
       1 300,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5052
  • HEWL122-5061
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,10 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 360,00 €
    1 360,00 €
    1 * 1 ST
       1 360,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5061
  • HEWL122-5062
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 360,00 €
    1 360,00 €
    1 * 1 ST
       1 360,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5062
  • HEWL122-5062E
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 520,00 €
    1 520,00 €
    1 * 1 ST
       1 520,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5062E
  • HEWL122-506E
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,50 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 360,00 €
    1 360,00 €
    1 * 1 ST
       1 360,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-506E
  • HEWL122-5063
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 360,00 €
    1 360,00 €
    1 * 1 ST
       1 360,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5063
  • HEWL122-5063E
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 520,00 €
    1 520,00 €
    1 * 1 ST
       1 520,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL122-5063E
  • HEWL123-500E
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,50 µm
    0,32 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 437,00 €
    437,00 €
    1 * 1 ST
       437,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-500E
  • HEWL123-500ELTM
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,50 µm
    0,32 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 640,00 €
    1 640,00 €
    1 * 1 ST
       1 640,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-500ELTM
  • HEWL123-5011
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,10 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 497,00 €
    497,00 €
    1 * 1 ST
       497,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5011
  • HEWL123-5011LTM
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    0,10 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 700,00 €
    1 700,00 €
    1 * 1 ST
       1 700,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5011LTM
  • HEWL123-5012
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 497,00 €
    497,00 €
    1 * 1 ST
       497,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5012
  • HEWL123-5012E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 558,00 €
    558,00 €
    1 * 1 ST
       558,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5012E
  • HEWL123-5012LTM
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 700,00 €
    1 700,00 €
    1 * 1 ST
       1 700,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5012LTM
  • HEWL123-5013
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 497,00 €
    497,00 €
    1 * 1 ST
       497,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5013
  • HEWL123-5013E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 558,00 €
    558,00 €
    1 * 1 ST
       558,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5013E
  • HEWL123-5013LTM
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 700,00 €
    1 700,00 €
    1 * 1 ST
       1 700,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5013LTM
  • HEWL123-502D
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,17 µm
    0,32 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 738,00 €
    738,00 €
    1 * 1 ST
       738,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-502D
  • HEWL123-5022
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,32 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 738,00 €
    738,00 €
    1 * 1 ST
       738,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5022
  • HEWL123-5026
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,52 µm
    0,32 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 738,00 €
    738,00 €
    1 * 1 ST
       738,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5026
  • HEWL123-502F
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,05 µm
    0,32 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 738,00 €
    738,00 €
    1 * 1 ST
       738,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-502F
  • HEWL123-5031
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,10 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 832,00 €
    832,00 €
    1 * 1 ST
       832,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5031
  • HEWL123-5032
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 832,00 €
    832,00 €
    1 * 1 ST
       832,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5032
  • HEWL123-5032E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 934,00 €
    934,00 €
    1 * 1 ST
       934,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5032E
  • HEWL123-5032LTM
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 080,00 €
    2 080,00 €
    1 * 1 ST
       2 080,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5032LTM
  • HEWL123-503E
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 832,00 €
    832,00 €
    1 * 1 ST
       832,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-503E
  • HEWL123-503ELTM
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    0,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 080,00 €
    2 080,00 €
    1 * 1 ST
       2 080,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-503ELTM
  • HEWL123-5033
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 832,00 €
    832,00 €
    1 * 1 ST
       832,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5033
  • HEWL123-5033E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 934,00 €
    934,00 €
    1 * 1 ST
       934,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5033E
  • HEWL123-503B
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 832,00 €
    832,00 €
    1 * 1 ST
       832,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-503B
  • HEWL123-503BLTM
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    1,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 080,00 €
    2 080,00 €
    1 * 1 ST
       2 080,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-503BLTM
  • HEWL123-5003LTM
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,32 mm
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 520,00 €
    1 520,00 €
    1 * 1 ST
       1 520,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5003LTM
  • HEWL123-5052
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,32 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 330,00 €
    1 330,00 €
    1 * 1 ST
       1 330,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5052
  • HEWL123-5056
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,52 µm
    0,32 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 330,00 €
    1 330,00 €
    1 * 1 ST
       1 330,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5056
  • HEWL123-5053
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,32 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 330,00 €
    1 330,00 €
    1 * 1 ST
       1 330,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5053
  • HEWL123-5062
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 480,00 €
    1 480,00 €
    1 * 1 ST
       1 480,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5062
  • HEWL123-5063
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 480,00 €
    1 480,00 €
    1 * 1 ST
       1 480,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL123-5063
  • HEWL124-5037
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,42 µm
    0,45 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL124-5037
  • HEWL124-5032
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,27 µm
    0,45 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL124-5032
  • HEWL125-50HB
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    2,65 µm
    0,53 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 423,00 €
    423,00 €
    1 * 1 ST
       423,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-50HB
  • HEWL125-501K
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 508,00 €
    508,00 €
    1 * 1 ST
       508,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-501K
  • HEWL125-5017
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 508,00 €
    508,00 €
    1 * 1 ST
       508,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5017
  • HEWL125-501J
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 508,00 €
    508,00 €
    1 * 1 ST
       508,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-501J
  • HEWL125-5012
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 508,00 €
    508,00 €
    1 * 1 ST
       508,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5012
  • HEWL125-5012E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    1,50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 570,00 €
    570,00 €
    1 * 1 ST
       570,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5012E
  • HEWL125-5025
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    5,00 µm
    0,53 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 842,00 €
    842,00 €
    1 * 1 ST
       842,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5025
  • HEWL125-503K
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,25 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-503K
  • HEWL125-5037
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5037
  • HEWL125-503D
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    0,88 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-503D
  • HEWL125-503J
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-503J
  • HEWL125-5032
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5032
  • HEWL125-503B
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    2,65 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-503B
  • HEWL125-5034
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5034
  • HEWL125-5035
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    5,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5035
  • HEWL125-5035E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    5,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5035E
  • HEWL125-5035LTM
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    5,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 190,00 €
    2 190,00 €
    1 * 1 ST
       2 190,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5035LTM
  • HEWL125-5032E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    1,50 µm
    0,53 mm
    32 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5032E
  • HEWL125-5032LTM
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    1,50 µm
    0,53 mm
    32 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 190,00 €
    2 190,00 €
    1 * 1 ST
       2 190,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5032LTM
  • HEWL125-5062
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    1,50 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 490,00 €
    1 490,00 €
    1 * 1 ST
       1 490,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5062
  • HEWL125-5065
    On 7" cage, high efficiency
    On 7" cage, high efficiency
    Capillary
    DB-5
    5,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 490,00 €
    1 490,00 €
    1 * 1 ST
       1 490,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5065
  • HEWL125-5065E
    On 5" cage, high efficiency
    On 5" cage, high efficiency
    Capillary
    DB-5
    5,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 670,00 €
    1 670,00 €
    1 * 1 ST
       1 670,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL125-5065E