Order Entry
Germany
ContactUsLinkComponent
Hazardous substance labels
Hazardous substance labels
Catalog # KROS27478
Supplier:  KROSCHKE
Hazardous substance labels
Catalog # KROS27478
Supplier:  KROSCHKE

Specifications

  • Colour:
    Black/red/white
  • Description:
    Hazard symbol question mark, foil
  • Dimensions:
    26×37 mm
  • Material:
    Foil
  • Pk:
    1 SET

Specifications

About this item

Hazard symbol label with question mark, used for unverified research material.

  • Self-adhesive
  • Resistant to water, alkalis and oil
  • Temperature resistant from –40 to +80 °C